Gene Transcripts

Gene Symbol
Dgcr6
Entrez Gene ID
303794
Gene Name
DiGeorge syndrome critical region gene 6
Gene Aliases
Chromosome Location
Chr. 11: 87076205 - 87081279 on Build Rnor_6.0
UniGene ID
Rn.22467
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107080.2 NP_001100550.1 4 509
XM_006248609.2 XP_006248671.1 4 547
GenBank mRNA BC158828.1 - 4 509

View Full Product Details