Gene Transcripts

Gene Symbol
Igf2bp2
Entrez Gene ID
303824
Gene Name
insulin-like growth factor 2 mRNA binding protein 2
Gene Aliases
RGD1305614
Chromosome Location
Chr. 11: 82466071 - 82621105 on Build Rnor_6.0
UniGene ID
Rn.17318
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270598.1 NP_001257527.1 9 1043
NM_001270599.1 NP_001257528.1 9 1043
GenBank mRNA FQ222567.1 - 9 811

View Full Product Details