Gene Transcripts

Gene Symbol
Atp13a5
Entrez Gene ID
303856
Gene Name
ATPase type 13A5
Gene Aliases
RGD1306792
Chromosome Location
Chr. 11: 74984546 - 75124249 on Build Rnor_6.0
UniGene ID
Rn.58246
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191657.1 NP_001178586.1 20 2455
XM_008768800.1 XP_008767022.1 20 2480

View Full Product Details