Gene Transcripts

Gene Symbol
Rwdd2b
Entrez Gene ID
304132
Gene Name
RWD domain containing 2B
Gene Aliases
RGD1305062
Chromosome Location
Chr. 11: 27085708 - 27094364 on Build Rnor_6.0
UniGene ID
Rn.206833
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100559.1 NP_001094029.1 2 211
GenBank mRNA BC161911.1 - 2 211

View Full Product Details