Gene Transcripts

Gene Symbol
Agfg2
Entrez Gene ID
304375
Gene Name
ArfGAP with FG repeats 2
Gene Aliases
Hrbl
Chromosome Location
Chr. 12: 22082041 - 22118239 on Build Rnor_6.0
UniGene ID
Rn.17775
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107131.1 NP_001100601.1 11 1386
XM_006249140.2 XP_006249202.1 12 1566
XM_006249141.2 XP_006249203.1 11 1532

View Full Product Details