Gene Transcripts

Gene Symbol
Alkbh2
Entrez Gene ID
304578
Gene Name
alkB, alkylation repair homolog 2 (E. coli)
Gene Aliases
RGD1306377
Chromosome Location
Chr. 12: 48257584 - 48262244 on Build Rnor_6.0
UniGene ID
Rn.107582
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126273.1 NP_001119745.1 3 898
XM_006249498.2 XP_006249560.1 4 637
GenBank mRNA BC166547.1 - 3 898

View Full Product Details