Gene Transcripts

Gene Symbol
Kctd3
Entrez Gene ID
305055
Gene Name
potassium channel tetramerization domain containing 3
Gene Aliases
Ren-45
Chromosome Location
Chr. 13: 107433588 - 107471843 on Build Rnor_6.0
UniGene ID
Rn.19263
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107199.1 NP_001100669.1 3 371
GenBank mRNA AY724501.1 - 3 360

View Full Product Details