Gene Transcripts

Gene Symbol
Il17rb
Entrez Gene ID
306247
Gene Name
interleukin 17 receptor B
Gene Aliases
Chromosome Location
Chr. 16: 6064287 - 6077978 on Build Rnor_6.0
UniGene ID
Rn.150863
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107290.1 NP_001100760.1 9 906
XM_006252603.2 XP_006252665.1 9 888
GenBank mRNA BC168886.1 - 5 451

View Full Product Details