Gene Transcripts

Gene Symbol
UBL4A
Entrez Gene ID
8266
Gene Name
ubiquitin like 4A
Gene Aliases
DX254E, DXS254E, G6PD, GDX, GET5, MDY2, TMA24, UBL4
Chromosome Location
Chr. X: 154483717 - 154486670 on Build GRCh38
UniGene ID
Hs.76480
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014235.4 NP_055050.1 3 283
GenBank mRNA AK000405.1 - 3 218
AK293227.1 - 2 445
BC043346.2 - 3 210 6058941
BC053589.1 - 3 244 5246329
DA012854.1 - 3 283
DR003955.1 - 3 223

View Full Product Details