Gene Transcripts

Gene Symbol
Fam50b
Entrez Gene ID
306883
Gene Name
family with sequence similarity 50, member B
Gene Aliases
RGD1563458
Chromosome Location
Chr. 17: 30864189 - 30883418 on Build Rnor_6.0
UniGene ID
Rn.59400
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107349.1 NP_001100819.1 2 1073
XM_006253827.2 XP_006253889.1 2 1427

View Full Product Details