Gene Transcripts
- Gene Symbol
- Btn2a2
- Entrez Gene ID
- 306957
- Gene Name
- butyrophilin, subfamily 2, member A2
- Gene Aliases
- Chromosome Location
- Chr. 17: 43867249 - 43878732 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_225366.8 |
XP_225366.6
|
4 |
983 |
|
View Full Product Details