Gene Transcripts

Gene Symbol
Hars2
Entrez Gene ID
307491
Gene Name
histidyl-tRNA synthetase 2, mitochondrial
Gene Aliases
Hars2l, RGD1308426, Zmat2
Chromosome Location
Chr. 18: 29629201 - 29638460 on Build Rnor_6.0
UniGene ID
Rn.139312
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014012.2 NP_001014034.2 12, 13 1581
XM_006254575.2 XP_006254637.1 11, 12 1491
GenBank mRNA BC089973.1 - 5, 6 745

View Full Product Details