Gene Transcripts

Gene Symbol
Fa2h
Entrez Gene ID
307855
Gene Name
fatty acid 2-hydroxylase
Gene Aliases
RGD1310347, Wdr59
Chromosome Location
Chr. 19: 43545380 - 43596788 on Build Rnor_6.0
UniGene ID
Rn.67886
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135583.1 NP_001129055.1 4 627
GenBank mRNA DQ339484.1 - 4 627

View Full Product Details