Gene Transcripts

Gene Symbol
HIST1H3G
Entrez Gene ID
8355
Gene Name
histone cluster 1, H3g
Gene Aliases
H3/h, H3FH
Chromosome Location
Chr. 6: 26270918 - 26271384 on Build GRCh38
UniGene ID
Hs.247813
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003534.2 NP_003525.1 1 247
GenBank mRNA BC012185.1 - 1 678
BC079835.1 - 1 252

View Full Product Details