Gene Transcripts

Gene Symbol
Samd4b
Entrez Gene ID
308473
Gene Name
sterile alpha motif domain containing 4B
Gene Aliases
RGD1305597
Chromosome Location
Chr. 1: 85324010 - 85362241 on Build Rnor_6.0
UniGene ID
Rn.40291
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107498.1 NP_001100968.1 1 266
XM_008759143.1 XP_008757365.1 2 389
GenBank mRNA BC090319.1 - 2 356

View Full Product Details