Gene Transcripts

Gene Symbol
Lins
Entrez Gene ID
308704
Gene Name
lines homolog (Drosophila)
Gene Aliases
Lins1, Lins2, RGD1308792
Chromosome Location
Chr. 1: 127599501 - 127625583 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006229357.2 XP_006229419.1 5 1143
XM_006229358.2 XP_006229420.1 4 937
XM_008759572.1 XP_008757794.1 5 1265
XM_008759573.1 XP_008757795.1 5 1224
XM_008759574.1 XP_008757796.1 4 1320

View Full Product Details