Gene Transcripts
- Gene Symbol
- OR1D5
- Entrez Gene ID
- 8386
- Gene Name
- olfactory receptor family 1 subfamily D member 5
- Gene Aliases
- C17orf2, OR17-2, OR17-30, OR17-31, OR1D4
- Chromosome Location
- Chr. 17: 3062669 - 3063607 on Build GRCh38
- UniGene ID
- Hs.553508
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014566.1 |
NP_055381.1
|
1 |
643 |
|
View Full Product Details