Gene Transcripts

Gene Symbol
OR1D5
Entrez Gene ID
8386
Gene Name
olfactory receptor family 1 subfamily D member 5
Gene Aliases
C17orf2, OR17-2, OR17-30, OR17-31, OR1D4
Chromosome Location
Chr. 17: 3062669 - 3063607 on Build GRCh38
UniGene ID
Hs.553508
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014566.1 NP_055381.1 1 522
GenBank mRNA BC106876.2 - 1 523

View Full Product Details