Gene Transcripts

Gene Symbol
Chrdl2
Entrez Gene ID
308854
Gene Name
chordin-like 2
Gene Aliases
Chromosome Location
Chr. 1: 165008997 - 165036431 on Build Rnor_6.0
UniGene ID
Rn.78674
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107537.1 NP_001101007.1 6 705

View Full Product Details