Gene Transcripts

Gene Symbol
Tm9sf3
Entrez Gene ID
309475
Gene Name
transmembrane 9 superfamily member 3
Gene Aliases
RGD1564625
Chromosome Location
Chr. 1: 260464935 - 260517997 on Build Rnor_6.0
UniGene ID
Rn.17150
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006231413.1 XP_006231475.1 4 898
GenBank mRNA FQ209899.1 - 1 13

View Full Product Details