Gene Transcripts

Gene Symbol
RASAL1
Entrez Gene ID
8437
Gene Name
RAS protein activator like 1
Gene Aliases
RASAL
Chromosome Location
Chr. 12: 113096515 - 113136239 on Build GRCh38
UniGene ID
Hs.528693
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193520.1 NP_001180449.1 17 2023
NM_001193521.1 NP_001180450.1 16 2008
NM_001301202.1 NP_001288131.1 16 1856
NM_004658.2 NP_004649.2 17 2032
XM_005253950.4 XP_005254007.1 17 2038
XM_006719641.3 XP_006719704.1 17 1877
XM_006719642.3 XP_006719705.1 16 2007
XM_011538852.2 XP_011537154.1 16 2010
XM_011538853.2 XP_011537155.1 16 2004
XM_011538854.2 XP_011537156.1 6 750
XM_017020028.1 XP_016875517.1 17 2035
XM_017020029.1 XP_016875518.1 16 2010
XM_017020030.1 XP_016875519.1 17 2038
XM_017020031.1 XP_016875520.1 17 2035
GenBank mRNA AB208935.1 - 17 2008
AF086713.1 - 17 2009
AK294350.1 - 17 2035
AL136672.1 - 16 1951
BC014420.1 - 16 1899 4638571
BC093724.1 - 16 1758 7939569
BC143261.1 - 16 1761 9051763

View Full Product Details