Gene Transcripts

Gene Symbol
Pamr1
Entrez Gene ID
311252
Gene Name
peptidase domain containing associated with muscle regeneration 1
Gene Aliases
RGD1308745, Ramp
Chromosome Location
Chr. 3: 92403532 - 92502094 on Build Rnor_6.0
UniGene ID
Rn.230588
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107755.1 NP_001101225.1 2 269
XM_008762061.1 XP_008760283.1 2 386

View Full Product Details