Gene Transcripts

Gene Symbol
Qser1
Entrez Gene ID
311266
Gene Name
glutamine and serine rich 1
Gene Aliases
RGD1311804
Chromosome Location
Chr. 3: 94702635 - 94767370 on Build Rnor_6.0
UniGene ID
Rn.123081
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001139493.1 NP_001132965.1 3 3847
XM_006234646.2 XP_006234708.1 3 3950

View Full Product Details