Gene Transcripts

Gene Symbol
Tox2
Entrez Gene ID
311615
Gene Name
TOX high mobility group box family member 2
Gene Aliases
GCX-1, Gcx1
Chromosome Location
Chr. 3: 159569329 - 159694772 on Build Rnor_6.0
UniGene ID
Rn.129021
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199392.2 NP_955424.2 4, 5 677
XM_006235578.2 XP_006235640.1 4, 5 711
GenBank mRNA AB096685.1 - 4, 5 678

View Full Product Details