Gene Transcripts

Gene Symbol
Cdh26
Entrez Gene ID
311693
Gene Name
cadherin 26
Gene Aliases
Chromosome Location
Chr. 3: 173969019 - 174020867 on Build Rnor_6.0
UniGene ID
Rn.228188
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191745.1 NP_001178674.1 5 500
XM_008762509.1 XP_008760731.1 5 754

View Full Product Details