Gene Transcripts

Gene Symbol
Slc27a4
Entrez Gene ID
311839
Gene Name
solute carrier family 27 (fatty acid transporter), member 4
Gene Aliases
Fatp4
Chromosome Location
Chr. 3: 8363937 - 8376858 on Build Rnor_6.0
UniGene ID
Rn.145068
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100706.1 NP_001094176.1 13 2160
GenBank mRNA BC091430.1 - 4 405

View Full Product Details