Gene Transcripts

Gene Symbol
Dennd1a
Entrez Gene ID
311913
Gene Name
DENN/MADD domain containing 1A
Gene Aliases
RGD1307927
Chromosome Location
Chr. 3: 22061120 - 22501492 on Build Rnor_6.0
UniGene ID
Rn.92793
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191747.1 NP_001178676.1 7, 8 505
XM_008761756.1 XP_008759978.1 7, 8 1232
XM_008761757.1 XP_008759979.1 7, 8 1232

View Full Product Details