Gene Transcripts

Gene Symbol
Wdr91
Entrez Gene ID
312225
Gene Name
WD repeat domain 91
Gene Aliases
RGD1306697
Chromosome Location
Chr. 4: 62398073 - 62438958 on Build Rnor_6.0
UniGene ID
Rn.158722
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127298.2 NP_001120770.2 1 116
GenBank mRNA BC166785.1 - 1 116

View Full Product Details