Gene Transcripts

Gene Symbol
C1r
Entrez Gene ID
312705
Gene Name
complement component 1, r subcomponent
Gene Aliases
Chromosome Location
Chr. 4: 157126060 - 157136825 on Build Rnor_6.0
UniGene ID
Rn.70397
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134555.1 NP_001128027.1 5 736
GenBank mRNA BC168673.1 - 5 736

View Full Product Details