Gene Transcripts

Gene Symbol
Xkr8
Entrez Gene ID
313033
Gene Name
XK, Kell blood group complex subunit-related family, member 8
Gene Aliases
RGD1305649, XRG8
Chromosome Location
Chr. 5: 150918600 - 150925727 on Build Rnor_6.0
UniGene ID
Rn.13596
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012099.1 NP_001012099.1 3 1983
GenBank mRNA AY534262.1 - 3 1983
BC105851.1 - 3 1110 7191341

View Full Product Details