Gene Transcripts

Gene Symbol
Klhl32
Entrez Gene ID
313111
Gene Name
kelch-like family member 32
Gene Aliases
RGD1310364
Chromosome Location
Chr. 5: 38972742 - 39215187 on Build Rnor_6.0
UniGene ID
Rn.26637
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_232840.8 XP_232840.4 9 1952
GenBank mRNA BC087711.1 - 10 1779

View Full Product Details