Gene Transcripts
- Gene Symbol
- Zyg11a
- Entrez Gene ID
- 313482
- Gene Name
- zyg-11 family member A, cell cycle regulator
- Gene Aliases
- RGD1309582
- Chromosome Location
- Chr. 5: 127790496 - 127828911 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_233348.8 |
XP_233348.6
|
3 |
655 |
|
View Full Product Details