Gene Transcripts

Gene Symbol
Zswim5
Entrez Gene ID
313524
Gene Name
zinc finger, SWIM-type containing 5
Gene Aliases
Chromosome Location
Chr. 5: 135735825 - 135854525 on Build Rnor_6.0
UniGene ID
Rn.134685
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107963.1 NP_001101433.1 13 2226
XM_008764009.1 XP_008762231.1 13 2148

View Full Product Details