Gene Transcripts

Gene Symbol
Dhrs3
Entrez Gene ID
313689
Gene Name
dehydrogenase/reductase (SDR family) member 3
Gene Aliases
Chromosome Location
Chr. 5: 162809090 - 162843385 on Build Rnor_6.0
UniGene ID
Rn.12011
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037199.3 NP_001032276.3 5, 6 1115
GenBank mRNA BC107450.1 - 5, 6 1115
EF125189.1 - 5, 6 1561

View Full Product Details