Gene Transcripts

Gene Symbol
Acap3
Entrez Gene ID
313772
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
Gene Aliases
Centb5
Chromosome Location
Chr. 5: 173340060 - 173354756 on Build Rnor_6.0
UniGene ID
Rn.144386
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107999.1 NP_001101469.1 21 2622
XM_006239577.2 XP_006239639.1 21 2158

View Full Product Details