Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001014078.1 | NP_001014100.1 | 2 | 1000 | |
XM_006240232.2 | XP_006240294.1 | 3 | 1299 | ||
XM_006240233.2 | XP_006240295.1 | 3 | 1039 | ||
XM_006240235.2 | XP_006240297.2 | 2 | 996 | ||
GenBank mRNA | BC089849.1 | - | 2 | 1000 |