Gene Transcripts

Gene Symbol
Macc1
Entrez Gene ID
314539
Gene Name
metastasis associated in colon cancer 1
Gene Aliases
7a5, RGD1309838
Chromosome Location
Chr. 6: 147315254 - 147392530 on Build Rnor_6.0
UniGene ID
Rn.81938
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191775.1 NP_001178704.1 2 1688
XM_006240694.2 XP_006240756.1 3 2027
XM_008765003.1 XP_008763225.1 4 2466

View Full Product Details