Gene Transcripts

Gene Symbol
AKR7A2
Entrez Gene ID
8574
Gene Name
aldo-keto reductase family 7 member A2
Gene Aliases
AFAR, AFAR1, AFB1-AR1, AKR7
Chromosome Location
Chr. 1: 19302708 - 19312146 on Build GRCh38
UniGene ID
Hs.571886
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320979.1 NP_001307908.1 6 1076
NM_003689.3 NP_003680.2 7 1181
GenBank mRNA AF026947.1 - 7 1149
BC004111.2 - 7 1108 3832140
BC007352.2 - 7 1156
BC010852.1 - 7 1157 3898226
BC011586.1 - 7 1108 4620484
BC012171.1 - 7 1156 4642811
BC013996.2 - 7 1112 3625337
BU944847.1 - 1 252
EU794591.1 - 7 1181
Y16675.1 - 7 1143

View Full Product Details