Gene Transcripts
- Gene Symbol
- Fam84b
- Entrez Gene ID
- 315019
- Gene Name
- family with sequence similarity 84, member B
- Gene Aliases
- RGD1562236
- Chromosome Location
- Chr. 7: 101138188 - 101142123 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_235349.8 |
XP_235349.1
|
2 |
1988 |
|
View Full Product Details