Gene Transcripts
- Gene Symbol
- Trappc9
- Entrez Gene ID
- 315059
- Gene Name
- trafficking protein particle complex 9
- Gene Aliases
- Nibp, RGD1309461
- Chromosome Location
- Chr. 7: 113986363 - 114309056 on Build Rnor_6.0
- UniGene ID
- Rn.155796
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
FM067737.1 |
- |
4 |
461 |
|
View Full Product Details