Gene Transcripts

Gene Symbol
Foxr1
Entrez Gene ID
315601
Gene Name
forkhead box R1
Gene Aliases
Chromosome Location
Chr. 8: 48754245 - 48762761 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006242968.2 XP_006243030.1 1, 2 484
XM_006242969.2 XP_006243031.1 1, 2 484

View Full Product Details