Gene Transcripts

Gene Symbol
Adpgk
Entrez Gene ID
315722
Gene Name
ADP-dependent glucokinase
Gene Aliases
RGD1306103
Chromosome Location
Chr. 8: 64088877 - 64116832 on Build Rnor_6.0
UniGene ID
Rn.66171
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100723.1 NP_001094193.1 3, 4 560
XM_006243170.2 XP_006243232.1 3, 4 596
GenBank mRNA BC090343.1 - 2, 3 267

View Full Product Details