Gene Transcripts

Gene Symbol
Fam81a
Entrez Gene ID
315789
Gene Name
family with sequence similarity 81, member A
Gene Aliases
RGD1311958
Chromosome Location
Chr. 8: 76521659 - 76579387 on Build Rnor_6.0
UniGene ID
Rn.163030
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108163.1 NP_001101633.1 5, 6 903
XM_006243378.2 XP_006243440.1 5, 6 739
XM_006243379.2 XP_006243441.1 5, 6 811

View Full Product Details