Gene Transcripts

Gene Symbol
Nek11
Entrez Gene ID
315978
Gene Name
NIMA-related kinase 11
Gene Aliases
Chromosome Location
Chr. 8: 113703406 - 113937097 on Build Rnor_6.0
UniGene ID
Rn.105331
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108182.1 NP_001101652.1 2 594
XM_008766530.1 XP_008764752.1 1 228

View Full Product Details