Gene Transcripts

Gene Symbol
Slc22a13
Entrez Gene ID
316062
Gene Name
solute carrier family 22 (organic anion/urate transporter), member 13
Gene Aliases
Chromosome Location
Chr. 8: 127885268 - 127900469 on Build Rnor_6.0
UniGene ID
Rn.99291
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126285.1 NP_001119757.1 3 598
XM_008766677.1 XP_008764899.1 3 604
XM_008766678.1 XP_008764900.1 3 604
GenBank mRNA BC166511.1 - 3 598

View Full Product Details