Gene Transcripts

Gene Symbol
Ccnyl1
Entrez Gene ID
316452
Gene Name
cyclin Y-like 1
Gene Aliases
RGD1310683
Chromosome Location
Chr. 9: 71398208 - 71434581 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008767164.1 XP_008765386.1 5 400
XM_008767165.1 XP_008765387.1 4 490
XM_008767166.1 XP_008765388.1 5 399

View Full Product Details