Gene Transcripts

Gene Symbol
Luc7l
Entrez Gene ID
360503
Gene Name
LUC7-like (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 10: 15538880 - 15572651 on Build Rnor_6.0
UniGene ID
Rn.99992
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024269.1 NP_001019440.1 5 798
XM_006246006.2 XP_006246068.1 6 763
XM_006246007.1 XP_006246069.1 6 2167
XM_008767588.1 XP_008765810.1 6 1626
GenBank mRNA BC089960.1 - 5 798

View Full Product Details