Gene Transcripts

Gene Symbol
Hmx1
Entrez Gene ID
360960
Gene Name
H6 family homeobox 1
Gene Aliases
Chromosome Location
Chr. 14: 80540558 - 80544613 on Build Rnor_6.0
UniGene ID
Rn.24574
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108363.1 NP_001101833.1 2 1335
XM_006251147.2 XP_006251209.1 2 1603

View Full Product Details