Gene Transcripts

Gene Symbol
Pck2
Entrez Gene ID
361042
Gene Name
phosphoenolpyruvate carboxykinase 2 (mitochondrial)
Gene Aliases
PEPCK-M
Chromosome Location
Chr. 15: 34216735 - 34224357 on Build Rnor_6.0
UniGene ID
Rn.35508
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108377.2 NP_001101847.2 9 1821
GenBank mRNA BC166766.1 - 9 1794

View Full Product Details