Gene Transcripts

Gene Symbol
Nsun2
Entrez Gene ID
361191
Gene Name
NOP2/Sun RNA methyltransferase family, member 2
Gene Aliases
RGD1311954
Chromosome Location
Chr. 1: 36295650 - 36319903 on Build Rnor_6.0
UniGene ID
Rn.27691
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108403.1 NP_001101873.1 2 223
XM_006227792.2 XP_006227854.1 2 230

View Full Product Details